
N-Methyl-N-(2-hydroxyethyl)-4-aminobenzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1201-91-8
:
Formula:C₁₀H₁₃NO₂
MDL Number:MFCD00963547
:179.216 g/mol
Melting Pt:66…70 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together