Order Entry
Italy
ContactUsLinkComponent
1,2,4,5-Tetrahydro-benzo[e][1,4]diazepin-3-one 95%
1,2,4,5-Tetrahydro-benzo[e][1,4]diazepin-3-one 95%
Catalog # APOSOR914172-1G
Supplier:  Apollo Scientific
CAS Number:  
1,2,4,5-Tetrahydro-benzo[e][1,4]diazepin-3-one 95%
Catalog # APOSOR914172-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :162.189 g/mol
Storage Temperature:Ambient
CAS Number:168080-43-1
Formula:C₉H₁₀N₂O
MDL Number:MFCD07784140