(R)-Ethyl piperidine-2-carboxylate hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:183786-20-1
:
Formula:C₈H₁₅NO₂·ClH
MDL Number:MFCD17926385
:193.671 g/mol
Storage Temperature:Ambient
Frequently Bought Together