Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 1-methyl-1H-imidazole-2-carboxylate
Methyl 1-methyl-1H-imidazole-2-carboxylate 95%
Methyl 1-methyl-1H-imidazole-2-carboxylate 95%
Methyl 1-methyl-1H-imidazole-2-carboxylate 95%
APOSOR910010-25G
:
Apollo Scientific
:
Methyl 1-methyl-1H-imidazole-2-carboxylate 95%
APOSOR910010-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
140.14 g/mol
:
Ambient
:
62366-53-4
:
C₆H₈N₂O₂
:
MFCD02179562