Order Entry
Italy
ContactUsLinkComponent
3(5)-Isopropyl-4-nitro-1H-pyrazole 98%
3(5)-Isopropyl-4-nitro-1H-pyrazole 98%
Catalog # APOSOR902381-1G
Supplier:  Apollo Scientific
CAS Number:  
3(5)-Isopropyl-4-nitro-1H-pyrazole 98%
Catalog # APOSOR902381-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :155.155 g/mol
Storage Temperature:Ambient
CAS Number:51355-77-2
Formula:C₆H₉N₃O₂
MDL Number:MFCD14657963