Order Entry
Italy
ContactUsLinkComponent
3-Methoxy-1-n,1-N-dimethylbenzene-1,4-diamine 95%
3-Methoxy-1-n,1-N-dimethylbenzene-1,4-diamine 95%
Catalog # APOSOR902314-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Methoxy-1-n,1-N-dimethylbenzene-1,4-diamine 95%
Catalog # APOSOR902314-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :166.22 g/mol
Storage Temperature:Ambient
CAS Number:7474-82-0
Formula:C₉H₁₄N₂O
MDL Number:MFCD16249531