Order Entry
Italy
ContactUsLinkComponent
2-Hydroxy-5-methoxypyridine 98%
2-Hydroxy-5-methoxypyridine 98%
Catalog # APOSOR901930-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Hydroxy-5-methoxypyridine 98%
Catalog # APOSOR901930-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :125.125 g/mol
Melting Pt:102…103 °C
Storage Temperature:Ambient
CAS Number:61941-79-5
Formula:C₆H₇NO₂
MDL Number:MFCD11044353