N-Methyl-2,3-dihydro-1,4-benzodioxine-6-carboxamide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:438026-68-7
:
Formula:C₁₀H₁₁NO₃
MDL Number:MFCD02713088
:193.199 g/mol
Storage Temperature:Ambient
Frequently Bought Together