Order Entry
Italy
ContactUsLinkComponent
3-(3-Chlorophenyl)propan-1-ol 96%
3-(3-Chlorophenyl)propan-1-ol 96%
Catalog # APOSOR900159-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(3-Chlorophenyl)propan-1-ol 96%
Catalog # APOSOR900159-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :170.636 g/mol
Storage Temperature:Ambient
CAS Number:22991-03-3
Formula:C₉H₁₁ClO
MDL Number:MFCD09028713