

[6-(Piperidin-1-yl)pyridin-2-yl]methylamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:868755-51-5
:
Formula:C₁₁H₁₇N₃
MDL Number:MFCD08690243
Boiling Pt:92 °C (0.1 mm Hg)
Storage Temperature:Ambient
UN:2735
ADR:8,III
:191.273 g/mol
:GHS05
:GHS07
Frequently Bought Together