

1-(Bromomethyl)pyrene
Catalog # APOSOR8551T-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:295.173 g/mol
Melting Pt:140…145 °C
:GHS05
:GHS07
Storage Temperature:Freezer
CAS Number:2595-90-6
Formula:C₁₇H₁₁Br
MDL Number:MFCD00269937