Order Entry
Italy
ContactUsLinkComponent
3-Pyridin-2-yl[1,2,3]triazolo[1,5-a]pyridine
Irritant
3-Pyridin-2-yl[1,2,3]triazolo[1,5-a]pyridine
Catalog # APOSOR8213-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Pyridin-2-yl[1,2,3]triazolo[1,5-a]pyridine
Catalog # APOSOR8213-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :196.208 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:947-88-6
Formula:C₁₁H₈N₄
MDL Number:MFCD08062393