Order Entry
Italy
ContactUsLinkComponent
4'-(Bromomethyl)-3'-chloro-[1,1'-biphenyl]-2-carbonitrile 98%
Irritant
4'-(Bromomethyl)-3'-chloro-[1,1'-biphenyl]-2-carbonitrile 98%
Catalog # APOSOR8211-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4'-(Bromomethyl)-3'-chloro-[1,1'-biphenyl]-2-carbonitrile 98%
Catalog # APOSOR8211-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :306.585 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:887268-24-8
Formula:C₁₄H₉BrClN
MDL Number:MFCD07777122