
2-Methyl-5-[(morpholin-4-yl)sulphonyl]-3-furoic acid 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:306936-37-8
:
Formula:C₁₀H₁₃NO₆S
MDL Number:MFCD02180756
:275.278 g/mol
Melting Pt:184 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together