

Methyl 5-(chlorosulphonyl)-1-methyl-1H-pyrrole-2-carboxylate
Catalog # APOSOR8129-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:237.661 g/mol
Melting Pt:79…80 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
:GHS07
CAS Number:306936-53-8
Formula:C₇H₈ClNO₄S
MDL Number:MFCD02180789