
4-(4-Methoxyphenyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine, tech
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:213462-19-2
:
Formula:C₁₄H₁₅NOS
MDL Number:MFCD00177096
:245.34 g/mol
Melting Pt:69…71 °C
Storage Temperature:Ambient
UN:3259
ADR:8,III
:GHS05
Frequently Bought Together