Order Entry
Italy
ContactUsLinkComponent
2-(2,4-Dinitrophenoxy)ethanol
Irritant
2-(2,4-Dinitrophenoxy)ethanol
Catalog # APOSOR7734-10G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(2,4-Dinitrophenoxy)ethanol
Catalog # APOSOR7734-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :228.159 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:2831-60-9
Formula:C₈H₈N₂O₆
MDL Number:MFCD00052808