Order Entry
Italy
ContactUsLinkComponent
1-(4'-Methoxy-[1,1'-biphenyl]-2-yl)ethan-1-one
Irritant
1-(4'-Methoxy-[1,1'-biphenyl]-2-yl)ethan-1-one
Catalog # APOSOR7471-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(4'-Methoxy-[1,1'-biphenyl]-2-yl)ethan-1-one
Catalog # APOSOR7471-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :226.27 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:192863-43-7
Formula:C₁₅H₁₄O₂
MDL Number:MFCD04039102