(3S)-3-Methylpiperazine hydrochloride, N1-BOC protected
Catalog # APOSOR70192-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:236.739 g/mol
Storage Temperature:Refrigerator
CAS Number:1353006-46-8
Formula:C₁₀H₂₀N₂O₂·ClH
MDL Number:MFCD11101275