Order Entry
Italy
ContactUsLinkComponent
4-Isopropylbenzene-1,3-diol
4-Isopropylbenzene-1,3-diol
Catalog # APOSOR70132-25G
Supplier:  Apollo Scientific
CAS Number:  
4-Isopropylbenzene-1,3-diol
Catalog # APOSOR70132-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :152.19 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
 :GHS05
 :GHS07
CAS Number:23504-03-2
Formula:C₉H₁₂O₂
MDL Number:MFCD05663778