Order Entry
Italy
ContactUsLinkComponent
N-Methyl-N-[3-(2-morpholin-4-ylethoxy)benzyl]amine 97%
N-Methyl-N-[3-(2-morpholin-4-ylethoxy)benzyl]amine 97%
Catalog # APOSOR6797-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Methyl-N-[3-(2-morpholin-4-ylethoxy)benzyl]amine 97%
Catalog # APOSOR6797-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :250.337 g/mol
Melting Pt:170…172 °C
Storage Temperature:Ambient
CAS Number:857284-10-7
Formula:C₁₄H₂₂N₂O₂
MDL Number:MFCD08060519