
3-(Morpholin-4-ylmethyl)benzoic acid hydrochloride hydrate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:857283-67-1
:
Formula:C₁₂H₁₅NO₃·ClH·H₂O
MDL Number:MFCD07772833
:275.729 g/mol
Melting Pt:247…251 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together