Order Entry
Italy
ContactUsLinkComponent
[2-(1H-Pyrazol-1-yl)phenyl]methylamine 95%
Corrosive
Irritant
[2-(1H-Pyrazol-1-yl)phenyl]methylamine 95%
Catalog # APOSOR6729-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
[2-(1H-Pyrazol-1-yl)phenyl]methylamine 95%
Catalog # APOSOR6729-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :173.214 g/mol
Boiling Pt:100 °C (0.5 mm Hg)
Storage Temperature:Ambient
UN:2735
ADR:8,III
 :GHS05
 :GHS07
CAS Number:449758-13-8
Formula:C₁₀H₁₁N₃
MDL Number:MFCD03086189