Order Entry
Italy
ContactUsLinkComponent
(3S)-3-(Isopropylamino)piperidine, N1-BOC protected
(3S)-3-(Isopropylamino)piperidine, N1-BOC protected
Catalog # APOSOR62007-1G
Supplier:  Apollo Scientific
CAS Number:  
(3S)-3-(Isopropylamino)piperidine, N1-BOC protected
Catalog # APOSOR62007-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :242.358 g/mol
Storage Temperature:Ambient
CAS Number:1163286-38-1
Formula:C₁₃H₂₆N₂O₂
MDL Number:MFCD19381682