Order Entry
Italy
ContactUsLinkComponent
3-Aminopyrrolidine, 3-BOC protected hydrochloride 95+%
3-Aminopyrrolidine, 3-BOC protected hydrochloride 95+%
Catalog # APOSOR61180-5G
Supplier:  Apollo Scientific
CAS Number:  
3-Aminopyrrolidine, 3-BOC protected hydrochloride 95+%
Catalog # APOSOR61180-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :222.712 g/mol
Storage Temperature:Ambient
CAS Number:1188263-72-0
Formula:C₉H₁₈N₂O₂·ClH
MDL Number:MFCD11101358