Order Entry
Italy
ContactUsLinkComponent
2-(4-Bromophenyl)-2-azaspiro[4.4]nonane-1,3-dione
2-(4-Bromophenyl)-2-azaspiro[4.4]nonane-1,3-dione
Catalog # APOSOR61157-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Bromophenyl)-2-azaspiro[4.4]nonane-1,3-dione
Catalog # APOSOR61157-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :308.17 g/mol
Storage Temperature:Ambient
CAS Number:61343-15-5
Formula:C₁₄H₁₄BrNO₂
MDL Number:MFCD01763484