
3-Chloro-2-(morpholin-4-yl)pyridine-4-boronic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:957061-04-0
:
Formula:C₉H₁₂BClN₂O₃
MDL Number:MFCD09258746
Melting Pt:120…125 °C
:242.467 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together