Order Entry
Italy
ContactUsLinkComponent
N-Methyl-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)methylamine 97%
N-Methyl-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)methylamine 97%
Catalog # APOSOR59992-1G
Supplier:  Apollo Scientific
CAS Number:  
N-Methyl-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)methylamine 97%
Catalog # APOSOR59992-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :201.268 g/mol
Storage Temperature:Ambient
CAS Number:1031843-22-7
Formula:C₁₂H₁₅N₃
MDL Number:MFCD11109316