Ethyl 4-(methylsulphanyl)phenylacetate 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14062-27-2
:
Formula:C₁₁H₁₄O₂S
MDL Number:MFCD09751249
:210.293 g/mol
Melting Pt:55…58 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together