Order Entry
Italy
ContactUsLinkComponent
2-Acetamido-2-deoxy-D-glucono-1,5-lactone
2-Acetamido-2-deoxy-D-glucono-1,5-lactone
Catalog # APOSOR5600T-25MG
Supplier:  Apollo Scientific
CAS Number:  
2-Acetamido-2-deoxy-D-glucono-1,5-lactone
Catalog # APOSOR5600T-25MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :219.192 g/mol
Melting Pt:168…170 °C
Storage Temperature:Freezer
CAS Number:19026-22-3
Formula:C₈H₁₃NO₆
MDL Number:MFCD00076154