Order Entry
Italy
ContactUsLinkComponent
N,1-Di-Boc-3-(4-bromophenyl)-3-azetidinamine 95%
Irritant
N,1-Di-Boc-3-(4-bromophenyl)-3-azetidinamine 95%
Catalog # APOSOR55649-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N,1-Di-Boc-3-(4-bromophenyl)-3-azetidinamine 95%
Catalog # APOSOR55649-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :427.333 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1864803-36-0
Formula:C₁₉H₂₇BrN₂O₄
MDL Number:MFCD31657914