
(R)-4-Methyl-2-(hydroxymethyl)morpholine 95%
Catalog # APOSOR55502-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:131.173 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1159598-35-2
Formula:C₆H₁₃NO₂
MDL Number:MFCD16294696