
Thiomorpholine, N-BOC protected
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:220655-09-4
:
Formula:C₉H₁₇NO₂S
MDL Number:MFCD14635788
:203.302 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together