Order Entry
Italy
ContactUsLinkComponent
5-Methoxyquinolin-2(1H)-one 95+%
Irritant
5-Methoxyquinolin-2(1H)-one 95+%
Catalog # APOSOR55141-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Methoxyquinolin-2(1H)-one 95+%
Catalog # APOSOR55141-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :175.184 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:160893-04-9
Formula:C₁₀H₉NO₂
MDL Number:MFCD11877926