5-(5-Chlorosulphonyl-2-ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:139756-22-2
:
Formula:C₁₇H₁₉ClN₄O₄S
MDL Number:MFCD09753597
:410.875 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
Frequently Bought Together