
4,5,6,7-Tetrahydro[1,3]thiazolo[5,4-c]pyridine hydrochloride
Catalog # APOSOR52323-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:176.667 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1241725-84-7
Formula:C₆H₈N₂S·ClH
MDL Number:MFCD08726044