Order Entry
Italy
ContactUsLinkComponent
(1S)-1-(1,8a-Dihydroimidazo[1,2-a]pyridin-7-yl)ethanamine
(1S)-1-(1,8a-Dihydroimidazo[1,2-a]pyridin-7-yl)ethanamine
Catalog # APOSOR52297-500MG
Supplier:  Apollo Scientific
CAS Number:  
(1S)-1-(1,8a-Dihydroimidazo[1,2-a]pyridin-7-yl)ethanamine
Catalog # APOSOR52297-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :163.22 g/mol
Storage Temperature:Ambient
Formula:C₉H₁₃N₃
MDL Number:MFCD29042487