
Methyl 4,4-dimethyl-2-oxocyclopentanecarboxylate
Catalog # APOSOR52274-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:170.206 g/mol
Boiling Pt:55…60 °C (2 mm Hg)
:GHS07
Storage Temperature:Ambient
CAS Number:60585-44-6
Formula:C₉H₁₄O₃
MDL Number:MFCD07776759