Order Entry
Italy
ContactUsLinkComponent
Ethyl 3-methoxy-4-oxopiperidine-1-carboxylate 97%
Irritant
Ethyl 3-methoxy-4-oxopiperidine-1-carboxylate 97%
Catalog # APOSOR52157-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 3-methoxy-4-oxopiperidine-1-carboxylate 97%
Catalog # APOSOR52157-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :201.22 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:83863-72-3
Formula:C₉H₁₅NO₄
MDL Number:MFCD02093811