
4'-Bromo-[1,1'-biphenyl]-4-carboxaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:50670-58-1
:
Formula:C₁₃H₉BrO
MDL Number:MFCD05980925
:261.114 g/mol
Melting Pt:136…142 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together