
2-Bromo-4-hydroxy-5-methoxypyridine
Catalog # APOSOR51781-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:204.021 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1344046-11-2
Formula:C₆H₆BrNO₂
MDL Number:MFCD20327975