3-Iodo-6-methyl-1H-pyrazolo[3,4-d]pyrimidine ≥98%
Catalog # APOSOR510262-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:260.035 g/mol
Storage Temperature:Ambient
CAS Number:1415559-47-5
Formula:C₆H₅IN₄