Order Entry
Italy
ContactUsLinkComponent
[2,2'-Bipyridine]-5,5'-dicarboxaldehyde ≥98%
[2,2'-Bipyridine]-5,5'-dicarboxaldehyde ≥98%
Catalog # APOSOR510237-100MG
Supplier:  Apollo Scientific
CAS Number:  
[2,2'-Bipyridine]-5,5'-dicarboxaldehyde ≥98%
Catalog # APOSOR510237-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :212.204 g/mol
Storage Temperature:Ambient
CAS Number:135822-72-9
Formula:C₁₂H₈N₂O₂
MDL Number:MFCD03093215