1-Oxa-7-azaspiro[4.5]decane hydrochloride ≥98%
Catalog # APOSOR510219-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:177.672 g/mol
Storage Temperature:Ambient
CAS Number:1415562-85-4
Formula:C₈H₁₅NO·ClH
MDL Number:MFCD22572788