Order Entry
Italy
ContactUsLinkComponent
(1R,2R,4R)-rel-1-Azabicyclo[2,2,1]heptane-2-carboxylic acid hydrochloride ≥98%
(1R,2R,4R)-rel-1-Azabicyclo[2,2,1]heptane-2-carboxylic acid hydrochloride ≥98%
Catalog # APOSOR510198-1G
Supplier:  Apollo Scientific
CAS Number:  
(1R,2R,4R)-rel-1-Azabicyclo[2,2,1]heptane-2-carboxylic acid hydrochloride ≥98%
Catalog # APOSOR510198-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.629 g/mol
Storage Temperature:Ambient
CAS Number:921755-46-6
Formula:C₇H₁₁NO₂·ClH
MDL Number:MFCD22572789