Order Entry
Italy
ContactUsLinkComponent
 
7-Bromopyrido[3,2-d]pyrimidine-2,4(1H,3H)-dione ≥98%
Catalog # APOSOR510186-100MG
Supplier:  Apollo Scientific
CAS Number:  
7-Bromopyrido[3,2-d]pyrimidine-2,4(1H,3H)-dione ≥98%
Catalog # APOSOR510186-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :242.03 g/mol
Storage Temperature:Ambient
CAS Number:1215074-37-5
Formula:C₇H₄BrN₃O₂
MDL Number:MFCD13250073