Order Entry
Italy
ContactUsLinkComponent
6-Chloropyrazolo[1,5-a]pyrimidine-3-carboxylic acid ≥98%
6-Chloropyrazolo[1,5-a]pyrimidine-3-carboxylic acid ≥98%
Catalog # APOSOR510184-100MG
Supplier:  Apollo Scientific
CAS Number:  
6-Chloropyrazolo[1,5-a]pyrimidine-3-carboxylic acid ≥98%
Catalog # APOSOR510184-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :197.579 g/mol
Storage Temperature:Ambient
CAS Number:1204926-24-8
Formula:C₇H₄ClN₃O₂
MDL Number:MFCD20660461