Order Entry
Italy
ContactUsLinkComponent
2-Chloro-4-methoxy-5H-pyrrolo[3,2-d]pyrimidine ≥98%
2-Chloro-4-methoxy-5H-pyrrolo[3,2-d]pyrimidine ≥98%
Catalog # APOSOR510171-100MG
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-4-methoxy-5H-pyrrolo[3,2-d]pyrimidine ≥98%
Catalog # APOSOR510171-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :183.595 g/mol
Storage Temperature:Ambient
CAS Number:1375301-68-0
Formula:C₇H₆ClN₃O
MDL Number:MFCD21605181