Order Entry
Italy
ContactUsLinkComponent
4,5-Dimethylpyridine-3-sulphonyl chloride hydrochloride ≥98%
4,5-Dimethylpyridine-3-sulphonyl chloride hydrochloride ≥98%
Catalog # APOSOR510164-100MG
Supplier:  Apollo Scientific
CAS Number:  
4,5-Dimethylpyridine-3-sulphonyl chloride hydrochloride ≥98%
Catalog # APOSOR510164-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :242.123 g/mol
Storage Temperature:Ambient
Formula:C₇H₈ClNO₂S·ClH
MDL Number:MFCD31630727