Order Entry
Italy
ContactUsLinkComponent
6,8-Dichloroimidazo[1,2-a]pyrazine ≥98%
6,8-Dichloroimidazo[1,2-a]pyrazine ≥98%
Catalog # APOSOR510150-100MG
Supplier:  Apollo Scientific
CAS Number:  
6,8-Dichloroimidazo[1,2-a]pyrazine ≥98%
Catalog # APOSOR510150-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :188.014 g/mol
Storage Temperature:Ambient
CAS Number:1245645-38-8
Formula:C₆H₃Cl₂N₃
MDL Number:MFCD18072707